Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mpdu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301711.1 | NP_001288640.1 | 4 | 486 | ||
NM_011900.4 | NP_036030.2 | 4 | 451 | |||
GenBank mRNA | AB024713.1 | - | 4 | 379 | ||
AK040432.1 | - | 4 | 390 | |||
AK080233.1 | - | 4 | 445 | |||
AK089426.1 | - | 4 | 393 | |||
AK172020.1 | - | 4 | 412 | |||
AK183533.1 | - | 2 | 148 | |||
BC026776.1 | - | 4 | 373 | 4159355 | ||
BC048871.1 | - | 4 | 379 | 5364408 | ||
CV561188.1 | - | 4 | 437 |