Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Olfr926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146815.1 | NP_667026.1 | 1 | 132 | ||
GenBank mRNA | BC120558.1 | - | 1 | 184 | 40129481 | |
BC125490.1 | - | 1 | 184 | 40130005 |