Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 4921524J17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025722.3 | NP_079998.1 | 2 | 262 | ||
GenBank mRNA | AK007050.1 | - | 2 | 129 | ||
AK014959.1 | - | 2 | 255 | |||
AK017512.1 | - | 2 | 244 | |||
AK169408.1 | - | 2 | 199 | |||
BC027027.1 | - | 2 | 217 | |||
BC040087.1 | - | 2 | 181 | 3490529 | ||
BC050139.1 | - | 2 | 188 | 30009011 | ||
CK333687.1 | - | 2 | 262 |