Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ssna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023464.2 | NP_075953.1 | 3 | 395 | ||
GenBank mRNA | AB041656.1 | - | 3 | 389 | ||
AK004482.1 | - | 1 | 274 | |||
AK088418.1 | - | 3 | 327 | |||
AK158412.1 | - | 512 | ||||
AK208309.1 | - | 2 | 87 | |||
BC023033.1 | - | 3 | 358 | |||
BC037081.1 | - | 3 | 372 | 5098653 | ||
BC048360.1 | - | 3 | 373 | 5067090 | ||
BY703613.1 | - | 3 | 396 |