Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ssna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023464.2 | NP_075953.1 | 1 | 98 | ||
GenBank mRNA | AB041656.1 | - | 1 | 92 | ||
AK003356.2 | - | 1 | 99 | |||
AK088418.1 | - | 1 | 30 | |||
AK158412.1 | - | 46 | ||||
BC037081.1 | - | 1 | 75 | 5098653 | ||
BC048360.1 | - | 1 | 76 | 5067090 | ||
BY703613.1 | - | 1 | 99 |