Search
Search

Colec11| Gene Symbol: Colec11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027866.1 | NP_082142.1 | 7 | 687 | ||
| GenBank mRNA | AK003121.1 | - | 7 | 687 | ||
| BC104380.1 | - | 7 | 658 | 40050136 | ||
| BC104381.1 | - | 7 | 676 | 40050143 | ||