Search
Search

Antxr2| Gene Symbol: Antxr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133738.1 | NP_598499.1 | 14 | 1567 | ||
| GenBank mRNA | AK155460.1 | - | 14 | 1650 | ||
| AK166124.1 | - | 1339 | ||||
| AK170655.1 | - | 14 | 1205 | |||
| AK171292.1 | - | 14 | 1242 | |||
| BC003908.1 | - | 5 | 357 | |||
| BC076595.1 | - | 14 | 1567 | 30649897 | ||