Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Snx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026998.3 | NP_081274.2 | 11 | 954 | ||
GenBank mRNA | AK008135.1 | - | 6 | 419 | ||
AK013158.1 | - | 11 | 955 | |||
AK030764.1 | - | 5 | 284 | |||
BC061028.1 | - | 12 | 995 | 6594585 |