Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Thap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199042.2 | NP_950243.1 | 2 | 511 | ||
GenBank mRNA | AK014788.1 | - | 2 | 490 | ||
BC038639.1 | - | 2 | 467 | 5118383 |