Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Uqcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160356.1 | NP_001153828.1 | 2 | 328 | ||
GenBank mRNA | AK131602.1 | - | 2 | 319 | ||
AK131892.1 | - | 2 | 292 | |||
AK132116.1 | - | 2 | 319 | |||
AY941793.1 | - | 2 | 319 | |||
BC029863.1 | - | 2 | 253 | 4460168 | ||
BQ174007.1 | - | 1 | 516 |