Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Atad3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_179203.3 | NP_849534.2 | 13 | 1389 | ||
GenBank mRNA | AF343079.1 | - | 13 | 1435 | ||
AK075921.1 | - | 13 | 1385 | |||
AK148974.1 | - | 13 | 1388 | |||
AK149702.1 | - | 13 | 1388 | |||
AK150037.1 | - | 13 | 1388 | |||
AK165724.1 | - | 13 | 1387 | |||
AK166099.1 | - | 13 | 1385 | |||
AK172038.1 | - | 13 | 1385 | |||
AK172350.1 | - | 13 | 1390 | |||
AK173141.1 | - | 13 | 1422 | |||
AK218475.1 | - | 3 | 142 | |||
BC023301.1 | - | 13 | 1326 | 3671787 | ||
BC058373.1 | - | 13 | 1373 | 6828037 | ||
BC060036.1 | - | 13 | 1321 | 6530155 |