Search
Search

Dync2h1| Gene Symbol: Dync2h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029851.2 | NP_084127.2 | 81 | 11826 | ||
| GenBank mRNA | AK014500.1 | - | 8 | 779 | ||
| AK047623.1 | - | 5 | 403 | |||
| AK173324.1 | - | 23 | 2497 | |||
| AY452067.1 | - | 24 | 2535 | |||
| DQ104402.1 | - | 81 | 11826 | |||