Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cldn16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053241.5 | NP_444471.1 | 5 | 941 | ||
GenBank mRNA | AB009016.1 | - | 5 | 603 | ||
AF323748.1 | - | 5 | 731 | |||
AK085268.1 | - | 5 | 942 | |||
AK085333.1 | - | 5 | 917 | |||
BC115574.2 | - | 6 | 720 | 40102178 | ||
BC115575.2 | - | 6 | 720 | 40102179 |