Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dok4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053246.3 | NP_444476.1 | 5, 6 | 742 | ||
GenBank mRNA | AF418207.1 | - | 4, 5 | 626 | ||
AK032352.1 | - | 5, 6 | 732 | |||
AK043670.1 | - | 4, 5 | 654 | |||
AK077612.1 | - | 5, 6 | 734 | |||
AK139363.1 | - | 4, 5 | 630 | |||
AK142621.1 | - | 4, 5 | 649 | |||
AK156485.1 | - | 5, 6 | 742 | |||
BC004705.1 | - | 5, 6 | 703 | 3499419 |