Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133724.3 | NP_598485.1 | 1 | 547 | ||
GenBank mRNA | AK011795.1 | - | 1 | 519 | ||
BC019169.1 | - | 1 | 537 | 5002867 | ||
CK129009.1 | - | 1 | 547 |