Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other C1QBP SIRNA Assays ›
          Gene Symbol:
          C1qbp
          Gene Name:
          complement component 1, q subcomponent binding protein
          Gene Aliases:
          AA407365, AA986492, D11Wsu182e, HABP1, P32, gC1qBP
          Chromosome Location:
          Chr.11: 70977846 - 70983026 on Build GRCm38
          UniGene ID:
          Mm.30049
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS273438
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: C1qbp
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007573.2 NP_031599.2 2 326
          GenBank mRNA AF300619.1 - 2 265
          AJ001101.1 - 2 271
          AK010601.1 - 2 264
          AK010746.1 - 2 290
          AK011868.1 - 2 317
          AK169764.1 - 2 309
          BC038075.1 - 2 310 3154276
          BY275303.1 - 2 327

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          immune system process
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          apoptotic process
          complement activation, classical pathway
          RNA splicing
          phosphatidylinositol 3-kinase signaling
          regulation of complement activation
          negative regulation of interferon-gamma production
          negative regulation of interleukin-12 production
          negative regulation of MDA-5 signaling pathway
          negative regulation of RIG-I signaling pathway
          ribosome biogenesis
          mature ribosome assembly
          positive regulation of apoptotic process
          innate immune response
          positive regulation of cell adhesion
          negative regulation of mRNA splicing, via spliceosome
          negative regulation of defense response to virus
          positive regulation of protein kinase B signaling
          positive regulation of mitochondrial translation
          positive regulation of neutrophil chemotaxis
          positive regulation of substrate adhesion-dependent cell spreading
          positive regulation of trophoblast cell migration
          positive regulation of dendritic cell chemotaxis
          complement component C1q binding
          transcription corepressor activity
          mRNA binding
          protein kinase C binding
          protein binding
          hyaluronic acid binding
          transcription factor binding
          kininogen binding
          adrenergic receptor binding
          mitochondrial ribosome binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0