Search
Search

Ccni| Gene Symbol: Ccni | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017367.3 | NP_059063.2 | 6 | 1172 | ||
| GenBank mRNA | AF005886.2 | - | 6 | 824 | ||
| AK050465.1 | - | 6 | 1173 | |||
| AK079476.1 | - | 6 | 1113 | |||
| AK152614.1 | - | 6 | 1131 | |||
| AK177017.1 | - | 1 | 74 | |||
| AK178822.1 | - | 1 | 55 | |||
| AK184350.1 | - | 1 | 74 | |||
| AK188314.1 | - | 1 | 45 | |||
| AK203516.1 | - | 1 | 74 | |||
| AK214983.1 | - | 1 | 33 | |||
| AK219418.1 | - | 1 | 74 | |||
| BC003290.1 | - | 6 | 705 | 3583418 | ||
| BC029328.1 | - | 6 | 705 | |||