Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161624.1 | NP_001155096.1 | 2, 3 | 1115 | ||
NM_009876.4 | NP_034006.3 | 2, 3 | 1077 | |||
GenBank mRNA | AK077844.1 | - | 2, 3 | 1026 | ||
AK132278.1 | - | 2, 3 | 1024 | |||
BC005412.1 | - | 2, 3 | 1014 | 3490380 | ||
U20553.1 | - | 2, 3 | 948 | |||
U22399.1 | - | 2, 3 | 1006 |