Search
Search

Cox6a1| Gene Symbol: Cox6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007748.3 | NP_031774.1 | 2, 3 | 251 | ||
| GenBank mRNA | AK002402.1 | - | 2, 3 | 253 | ||
| AK019318.1 | - | 2, 3 | 256 | |||
| AK166496.1 | - | 2, 3 | 253 | |||
| BC003807.1 | - | 2, 3 | 251 | 3487598 | ||
| BC052816.1 | - | 2, 3 | 257 | 6439987 | ||
| L06465.1 | - | 2, 3 | 250 | |||
| U08440.1 | - | 2, 3 | 240 | |||