Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CSF1 SIRNA Assays ›
          Gene Symbol:
          Csf1
          Gene Name:
          colony stimulating factor 1 (macrophage)
          Gene Aliases:
          C87615, Csfm, MCSF, op
          Chromosome Location:
          Chr.3: 107741048 - 107760469 on Build GRCm38
          UniGene ID:
          Mm.795
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS273849
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Csf1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001113529.1 NP_001107001.1 8 1165
          NM_001113530.1 NP_001107002.1 8 2050
          NM_007778.4 NP_031804.3 8 2050
          GenBank mRNA AK138489.1 - 8 1824
          AK154261.1 - 8 2051
          AK154872.1 - 8 1824
          AK160995.1 - 8 1824
          AK170154.1 - 8 1760
          BC025593.1 - 8 951
          BC066187.1 - 8 1804 30106218
          BC066200.1 - 8 1804 30100524
          BC066205.1 - 8 1804 30100601
          M21149.1 - 8 1713
          M21952.1 - 8 1803
          S78392.1 - 8 918
          X05010.1 - 8 1825

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          positive regulation of cell-matrix adhesion
          osteoclast proliferation
          immune system process
          developmental process involved in reproduction
          inflammatory response
          transmembrane receptor protein tyrosine kinase signaling pathway
          cell proliferation
          positive regulation of cell proliferation
          positive regulation of gene expression
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of macrophage chemotaxis
          macrophage differentiation
          regulation of ossification
          osteoclast differentiation
          positive regulation of cell migration
          positive regulation of cellular protein metabolic process
          positive regulation of mononuclear cell proliferation
          macrophage colony-stimulating factor signaling pathway
          positive regulation of multicellular organism growth
          odontogenesis
          positive regulation of odontogenesis of dentin-containing tooth
          innate immune response
          positive regulation of macrophage differentiation
          positive regulation of monocyte differentiation
          positive regulation of osteoclast differentiation
          positive regulation of protein kinase activity
          positive regulation of Ras protein signal transduction
          homeostasis of number of cells within a tissue
          branching involved in mammary gland duct morphogenesis
          mammary gland fat development
          mammary duct terminal end bud growth
          positive regulation of macrophage colony-stimulating factor signaling pathway
          positive regulation of microglial cell migration
          cytokine activity
          macrophage colony-stimulating factor receptor binding
          protein binding
          growth factor activity
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0