Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DLX2 SIRNA Assays ›
          Gene Symbol:
          Dlx2
          Gene Name:
          distal-less homeobox 2
          Gene Aliases:
          AW121999, Dlx-2, Tes-1
          Chromosome Location:
          Chr.2: 71543408 - 71546754 on Build GRCm38
          UniGene ID:
          Mm.3896
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS273987
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Dlx2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010054.2 NP_034184.1 2 819
          GenBank mRNA AK144652.1 - 2 820
          BC094317.1 - 2 519 5718422
          M80540.1 - 2 757
          M83184.1 - 2 382

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_004405, L07919, AK316129, AB208823, AK291367, and others.
          This siRNA also targets the following transcripts in Rat: NM_001191746.

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          multicellular organism development
          proximal/distal pattern formation
          subpallium development
          hippocampus development
          olfactory bulb development
          forebrain neuron differentiation
          regulation of transcription from RNA polymerase II promoter involved in forebrain neuron fate commitment
          cerebral cortex GABAergic interneuron differentiation
          cerebral cortex GABAergic interneuron fate commitment
          cell differentiation
          odontogenesis of dentin-containing tooth
          negative regulation of Notch signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          embryonic cranial skeleton morphogenesis
          embryonic skeletal system development
          negative regulation of oligodendrocyte differentiation
          branching morphogenesis of a nerve
          cartilage development
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          chromatin binding
          single-stranded RNA binding
          protein binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0