Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DNM2 SIRNA Assays ›
          Gene Symbol:
          Dnm2
          Gene Name:
          dynamin 2
          Gene Aliases:
          Dyn2, Udnm, b2b2159Clo
          Chromosome Location:
          Chr.9: 21424908 - 21507759 on Build GRCm38
          UniGene ID:
          Mm.433257
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS274009
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Dnm2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039520.2 NP_001034609.1 20 2688
          NM_001253893.1 NP_001240822.1 20 2688
          NM_001253894.1 NP_001240823.1 20 2688
          NM_007871.2 NP_031897.2 19 2676
          GenBank mRNA AK005012.2 - 20 2664
          AK150046.1 - 20 2651
          AK154375.1 - 18 2338
          AK157844.1 - 20 2657
          AK170574.1 - 20 2664
          AK171049.1 - 20 2642
          AK171566.1 - 20 2658
          AK172231.1 - 20 2669
          AK190679.1 - 1 67
          AK212774.1 - 1 67
          BC010233.1 - 11 1404 3499181
          BY549643.1 - 1 14
          CD540553.1 - 2 375
          L31398.1 - 19 2616

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor internalization
          Golgi to plasma membrane transport
          endocytosis
          receptor-mediated endocytosis
          positive regulation of lamellipodium assembly
          negative regulation of transforming growth factor beta receptor signaling pathway
          regulation of axon extension
          receptor internalization
          transferrin transport
          regulation of Rac protein signal transduction
          macropinocytosis
          positive regulation of nitric oxide biosynthetic process
          positive regulation of endocytosis
          neuron projection morphogenesis
          positive regulation of phagocytosis
          cilium morphogenesis
          positive regulation of substrate adhesion-dependent cell spreading
          cellular response to dopamine
          regulation of Golgi organization
          positive regulation of sodium:potassium-exchanging ATPase activity
          negative regulation of membrane tubulation
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          hydrolase activity
          SH3 domain binding
          protein kinase binding
          D2 dopamine receptor binding
          protein complex binding
          phosphatidylinositol 3-kinase regulatory subunit binding
          WW domain binding
          nitric-oxide synthase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0