Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EEF1A1 SIRNA Assays ›
          Gene Symbol:
          Eef1a1 Gm6548
          Gene Name:
          eukaryotic translation elongation factor 1 alpha 1
          eukaryotic translation elongation factor 1 alpha 1 pseudogene
          Gene Aliases:
          -
          EG625054
          Chromosome Location:
          Chr.9: 78478453 - 78481724 on Build GRCm38
          Chromosome Location:
          Chr.17: 78850504 - 78852544 on Build GRCm38
          UniGene ID:
          Mm.138471
          Mm.311918
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID MSS274074
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Eef1a1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010106.2 NP_034236.2 3 297
          GenBank mRNA AK003188.1 - 3 245
          AK011771.1 - 3 244
          AK012341.1 - 3 245
          AK032914.1 - 3 243
          AK050635.1 - 3 244
          AK076696.1 - 3 245
          AK081725.1 - 3 245
          AK083361.1 - 3 245
          AK088041.1 - 3 245
          AK133725.1 - 3 245
          AK135448.1 - 3 246
          AK137223.1 - 3 245
          AK141243.1 - 3 245
          AK150798.1 - 3 245
          AK151479.1 - 3 246
          AK151773.1 - 3 245
          AK152810.1 - 3 245
          AK152993.1 - 3 245
          AK153387.1 - 3 245
          AK153402.1 - 3 245
          AK153851.1 - 3 245
          AK162049.1 - 3 245
          AK162313.1 - 3 244
          AK162440.1 - 3 249
          AK163799.1 - 3 245
          AK164113.1 - 3 247
          AK164671.1 - 3 245
          AK165938.1 - 3 245
          AK166021.1 - 3 245
          AK166033.1 - 3 245
          AK166058.1 - 3 245
          AK166089.1 - 3 245
          AK166615.1 - 3 245
          AK166807.1 - 3 245
          AK167843.1 - 3 245
          AK167873.1 - 3 244
          AK168424.1 - 3 245
          AK168467.1 - 3 251
          BC003969.1 - 2 215
          BC004005.1 - 2 206 3492770
          BC004067.1 - 2 234 3590965
          BC005660.1 - 2 235 3588966
          BC018223.1 - 3 233 4012653
          BC018485.1 - 3 247 3491827
          BC050124.1 - 1790
          BC068258.1 - 2042
          BC083069.1 - 3 233 6310448
          BC092053.1 - 2 223 4008574
          BC092276.1 - 2 228 3490007
          BC092292.1 - 2 189
          BC095965.1 - 2 223 6310404
          BC108391.1 - 3 230 30242277
          BY227095.1 - 3 274
          M22432.1 - 3 236
          X03688.1 - 2 235
          X13661.1 - 2 216
          Gene Symbol: Gm6548
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC089461.1 - 1 539

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          translation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          translational elongation
          protein homooligomerization
          cellular response to epidermal growth factor stimulus
          tRNA binding
          nucleotide binding
          mRNA binding
          translation elongation factor activity
          GTPase activity
          protein binding
          GTP binding
          protein kinase binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-6cc6g:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0