Search
Search

Il25| Gene Symbol: Il25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080729.3 | NP_542767.1 | 3 | 457 | ||
| GenBank mRNA | AF458060.1 | - | 2 | 381 | ||
| AY034088.1 | - | 2 | 333 | |||
| BC120568.1 | - | 3 | 456 | 40129491 | ||
| BC125524.1 | - | 3 | 456 | 40130039 | ||
| BC144962.1 | - | 3 | 456 | 9053510 | ||
| BX517868.1 | - | 2 | 266 | |||