Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GAL SIRNA Assays ›
Gene Symbol:
Gal
Gene Name:
galanin
Gene Aliases:
Galn
Chromosome Location:
Chr.19: 3409917 - 3414457 on Build GRCm38
UniGene ID:
Mm.4655
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS274463
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Contact Us
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Gal
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_010253.3
NP_034383.1
5
434
GenBank mRNA
AK133522.1
-
5
450
BC044055.1
-
5
434
6476459
Z23069.1
-
3
213
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_033237, J03624, M18102.
Gene Ontology Categories:
Function(s)
Process(es)
inflammatory response
neuropeptide signaling pathway
nervous system development
feeding behavior
negative regulation of cell proliferation
protein kinase A signaling
cAMP-mediated signaling
regulation of glucocorticoid metabolic process
positive regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
negative regulation of lymphocyte proliferation
positive regulation of cortisol secretion
positive regulation of catagen
positive regulation of large conductance calcium-activated potassium channel activity
negative regulation of root hair elongation
hormone activity
neuropeptide hormone activity
type 1 galanin receptor binding
type 2 galanin receptor binding
type 3 galanin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7wpsm:80/100.66.77.28:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline