Search
Search

Galk1| Gene Symbol: Galk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016905.2 | NP_058601.2 | 2 | 354 | ||
| GenBank mRNA | AB027012.1 | - | 2 | 310 | ||
| AK010340.1 | - | 2 | 351 | |||
| AK011103.1 | - | 2 | 355 | |||
| AK045694.1 | - | 2 | 328 | |||
| AK088151.1 | - | 2 | 355 | |||
| AK168716.1 | - | 2 | 314 | |||
| BC016602.1 | - | 2 | 354 | 3490224 | ||
| BC050151.1 | - | 2 | 325 | 30024030 | ||