Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Hist1h1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145713.4 | NP_663759.3 | 1 | 654 | ||
GenBank mRNA | AK054269.1 | - | 652 | |||
AK155411.1 | - | 654 | ||||
BC117045.1 | - | 1 | 651 | 40126364 | ||
BC119173.1 | - | 1 | 651 | 8733922 |
Gene Symbol: E430020m11rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK088581.1 | - | 1 | 449 |