Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008258.1 | NP_032284.1 | 5 | 437 | ||
GenBank mRNA | AK004909.1 | - | 5 | 452 | ||
AK010057.1 | - | 5 | 452 | |||
AK013122.1 | - | 5 | 449 | |||
AK014212.1 | - | 5 | 453 | |||
AK150410.1 | - | 5 | 456 | |||
AK152560.1 | - | 5 | 454 | |||
AK159784.1 | - | 5 | 451 | |||
AK167466.1 | - | 5 | 451 | |||
AK167961.1 | - | 5 | 441 | |||
AK168632.1 | - | 5 | 452 | |||
BC003758.1 | - | 5 | 426 | 3591830 | ||
U90123.1 | - | 5 | 437 |