Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LEP SIRNA Assays ›
          Gene Symbol:
          Lep
          Gene Name:
          leptin
          Gene Aliases:
          ob, obese
          Chromosome Location:
          Chr.6: 29060195 - 29073877 on Build GRCm38
          UniGene ID:
          Mm.277072
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS275329
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Lep
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008493.3 NP_032519.1 2 179
          GenBank mRNA AK030984.1 - 2 179
          AK142589.1 - 2 182
          BC125245.1 - 1 150 40054245
          FJ374142.1 - 1 120
          HQ166716.1 - 2 179
          U18812.1 - 2 176

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_013076, XM_008762762, D49653, D45862, U48849, and others.

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ovulation from ovarian follicle
          placenta development
          regulation of protein phosphorylation
          response to dietary excess
          cardiac muscle hypertrophy
          glucose metabolic process
          regulation of gluconeogenesis
          energy reserve metabolic process
          glycerol biosynthetic process
          lipid metabolic process
          fatty acid beta-oxidation
          signal transduction
          tyrosine phosphorylation of STAT protein
          cholesterol metabolic process
          bile acid metabolic process
          regulation of blood pressure
          positive regulation of cell proliferation
          adult feeding behavior
          fatty acid catabolic process
          negative regulation of metabolic process
          regulation of metabolic process
          central nervous system neuron development
          insulin secretion
          regulation of intestinal cholesterol absorption
          negative regulation of appetite
          response to insulin
          leptin-mediated signaling pathway
          positive regulation of peroxisome proliferator activated receptor signaling pathway
          intracellular signal transduction
          bone mineralization involved in bone maturation
          positive regulation of protein import into nucleus
          hormone metabolic process
          positive regulation of tyrosine phosphorylation of Stat3 protein
          glucose homeostasis
          eating behavior
          negative regulation of apoptotic process
          positive regulation of ion transport
          positive regulation of MAPK cascade
          regulation of fat cell differentiation
          positive regulation of myeloid cell differentiation
          negative regulation of vasoconstriction
          positive regulation of JAK-STAT cascade
          positive regulation of insulin receptor signaling pathway
          positive regulation of developmental growth
          regulation of insulin secretion
          regulation of steroid biosynthetic process
          leukocyte tethering or rolling
          regulation of lipoprotein lipid oxidation
          adipose tissue development
          negative regulation of cartilage development
          negative regulation of glucagon secretion
          regulation of protein localization to nucleus
          positive regulation of STAT protein import into nucleus
          positive regulation of reactive oxygen species metabolic process
          negative regulation of glutamine transport
          positive regulation of hepatic stellate cell activation
          hormone activity
          protein binding
          growth factor activity
          peptide hormone receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0