Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161769.1 | NP_001155241.1 | 4 | 590 | ||
NM_001161770.1 | NP_001155242.1 | 3 | 722 | |||
NM_010723.3 | NP_034853.1 | 4 | 877 | |||
GenBank mRNA | AF074600.1 | - | 3 | 438 | ||
AF096996.1 | - | 4 | 606 | |||
AF102817.1 | - | 4 | 507 | |||
AK080534.1 | - | 4 | 525 | |||
AK138278.1 | - | 3 | 723 | |||
AK159590.1 | - | 4 | 804 | |||
BC003488.1 | - | 3 | 589 | 2811636 | ||
BC004661.1 | - | 4 | 530 | |||
BC010278.1 | - | 4 | 620 | 3589532 | ||
BC100382.1 | - | 4 | 482 | 6487478 |