Search
Search

Nudt1| Gene Symbol: Nudt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008637.1 | NP_032663.1 | 3 | 116 | ||
| GenBank mRNA | AK011695.1 | - | 3 | 105 | ||
| AK088309.1 | - | 3 | 108 | |||
| AK168312.1 | - | 3 | 119 | |||
| BC021940.1 | - | 2 | 87 | 5067377 | ||
| BC098239.1 | - | 2 | 86 | 3469785 | ||
| D49956.1 | - | 3 | 116 | |||