This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
Olfr16
- Gene Name:
-
olfactory receptor 16
- Gene Aliases:
-
MOR23, MOR26713
- Chromosome Location:
-
Chr.1: 172956768 - 172957817 on Build GRCm38
- UniGene ID:
-
Mm.377103
- Species:
- Mouse
- Species Specific ID (Flybase ID):
-
-
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System