Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Abcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008830.2 | NP_032856.2 | 13 | 1560 | ||
GenBank mRNA | AK050345.1 | - | 12 | 1513 | ||
BC150687.1 | - | 13 | 1430 | 9087598 | ||
J03398.1 | - | 13 | 1560 |