Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PSAP SIRNA Assays ›
Gene Symbol:
Psap
Gene Name:
prosaposin
Gene Aliases:
AI037048, SGP-1
Chromosome Location:
Chr.10: 60277628 - 60302600 on Build GRCm38
UniGene ID:
Mm.277498
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS276490
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Contact Us
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Psap
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146120.1
NP_001139592.1
6
747
NM_001146121.1
NP_001139593.1
6
747
NM_001146122.1
NP_001139594.1
6
747
NM_001146123.1
NP_001139595.1
6
747
NM_001146124.1
NP_001139596.1
6
663
NM_011179.3
NP_035309.3
6
747
GenBank mRNA
AK088369.1
-
6
661
AK089998.1
-
6
665
AK093881.1
-
6
661
AK135637.1
-
6
661
AK145607.1
-
6
668
AK145802.1
-
6
662
AK148150.1
-
6
669
AK148152.1
-
6
688
AK148545.1
-
6
663
AK149785.1
-
6
664
AK150331.1
-
6
663
AK150645.1
-
6
661
AK151034.1
-
6
661
AK151173.1
-
6
662
AK151230.1
-
6
661
AK151234.1
-
6
663
AK151251.1
-
6
661
AK151525.1
-
6
663
AK151529.1
-
6
663
AK151686.1
-
6
697
AK151852.1
-
6
663
AK151875.1
-
6
660
AK151891.1
-
6
661
AK151907.1
-
6
663
AK151943.1
-
6
663
AK152117.1
-
6
663
AK152126.1
-
6
661
AK152134.1
-
6
663
AK152203.1
-
6
695
AK152280.1
-
6
661
AK152319.1
-
6
665
AK152355.1
-
6
663
AK152408.1
-
6
666
AK152505.1
-
6
663
AK152557.1
-
6
663
AK152605.1
-
6
663
AK152819.1
-
6
675
AK153404.1
-
6
661
AK159108.1
-
6
691
AK159164.1
-
6
661
AK159167.1
-
6
662
AK159198.1
-
6
661
AK159221.1
-
6
665
AK159244.1
-
6
661
AK159266.1
-
6
661
AK159333.1
-
6
661
AK159341.1
-
6
661
AK159384.1
-
6
661
AK159414.1
-
6
664
AK159427.1
-
6
663
AK159451.1
-
6
661
AK159457.1
-
6
675
AK159463.1
-
6
663
AK159487.1
-
6
661
AK159492.1
-
6
661
AK159510.1
-
6
661
AK159528.1
-
6
661
AK159530.1
-
6
661
AK159589.1
-
6
663
AK159604.1
-
6
661
AK159622.1
-
5
553
AK159634.1
-
6
661
AK159638.1
-
6
662
AK159657.1
-
6
661
AK159664.1
-
6
661
AK159685.1
-
6
661
AK159692.1
-
6
661
AK159709.1
-
6
661
AK159720.1
-
6
661
AK159722.1
-
6
661
AK159728.1
-
6
663
AK159840.1
-
6
661
AK159861.1
-
6
661
AK159862.1
-
6
663
AK159877.1
-
6
661
AK159910.1
-
6
661
AK159915.1
-
6
661
AK159936.1
-
6
663
AK159982.1
-
6
661
AK159994.1
-
6
661
AK160015.1
-
6
661
AK160019.1
-
6
663
AK160021.1
-
6
661
AK160024.1
-
6
661
AK160046.1
-
6
661
AK160049.1
-
6
663
AK160073.1
-
6
661
AK160108.1
-
6
661
AK160877.1
-
6
664
AK161313.1
-
6
678
AK166700.1
-
6
661
AK166707.1
-
6
660
AK166744.1
-
6
659
AK166811.1
-
6
661
AK166849.1
-
6
661
AK166879.1
-
6
663
AK166909.1
-
6
661
AK166920.1
-
6
661
AK166946.1
-
6
666
AK166959.1
-
6
661
AK166984.1
-
6
661
AK167014.1
-
6
661
AK167025.1
-
6
661
AK167073.1
-
6
661
AK167099.1
-
6
664
AK167108.1
-
6
661
AK167111.1
-
6
661
AK167147.1
-
6
661
AK167198.1
-
6
668
AK167213.1
-
6
661
AK167237.1
-
6
661
AK167243.1
-
6
662
AK167252.1
-
6
663
AK167418.1
-
6
663
AK167719.1
-
6
663
AK167902.1
-
6
731
AK170416.1
-
6
660
AK186105.1
-
1
48
AK189505.1
-
1
48
AK197814.1
-
1
48
BC036266.1
-
5
527
BC141089.1
-
6
660
9055701
BC145359.1
-
6
660
9053917
S36200.1
-
6
637
S71616.1
-
6
677
U27340.1
-
6
661
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
sphingolipid metabolic process
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
regulation of lipid metabolic process
regulation of MAPK cascade
positive regulation of MAPK cascade
developmental growth
prostate gland growth
epithelial cell differentiation involved in prostate gland development
cellular response to organic substance
negative regulation of hydrogen peroxide-induced cell death
G-protein coupled receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline