Search
Search

Pvalb| Gene Symbol: Pvalb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013645.3 | NP_038673.2 | 1, 2 | 90 | ||
| GenBank mRNA | AK002354.1 | - | 1, 2 | 98 | ||
| AK005371.1 | - | 1, 2 | 88 | |||
| AK013561.1 | - | 1, 2 | 82 | |||
| AK131986.1 | - | 1, 2 | 160 | |||
| BC027424.1 | - | 1, 2 | 90 | 4925213 | ||
| S75909.1 | - | 2, 3 | 151 | |||
| S75910.1 | - | 1027 | ||||
| X54613.1 | - | 1, 2 | 68 | |||
| X59382.1 | - | 2, 3 | 124 | |||