Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rfc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020022.2 | NP_064406.1 | 2 | 218 | ||
GenBank mRNA | AK011353.1 | - | 2 | 134 | ||
AK011649.1 | - | 2 | 132 | |||
AK075997.1 | - | 2 | 130 | |||
AK154681.1 | - | 2 | 172 | |||
BC023028.1 | - | 2 | 139 | 5374851 | ||
BC099370.1 | - | 2 | 182 | 30937690 | ||
BY338186.1 | - | 2 | 218 |