Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sar1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009120.2 | NP_033146.1 | 2 | 123 | ||
GenBank mRNA | AK157843.1 | - | 2 | 135 | ||
AK159239.1 | - | 2 | 134 | |||
AK168175.1 | - | 2 | 135 | |||
BC005549.1 | - | 2 | 124 | 3585667 | ||
L20294.1 | - | 2 | 98 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK129305.1 | - | 3321 | Chr. - - [-] on Build |