Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc12a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011390.2 | NP_035520.1 | 22 | 3053 | ||
GenBank mRNA | AF087436.1 | - | 22 | 3055 | ||
AK131129.1 | - | 22 | 3007 | |||
AK143535.1 | - | 22 | 3047 | |||
AK149750.1 | - | 22 | 3048 | |||
BC059242.1 | - | 22 | 3045 | |||
BC060133.1 | - | 22 | 3045 | |||
BC079677.1 | - | 18 | 4910 | |||
BC141107.1 | - | 22 | 3013 | 9055719 |