Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ints9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253731.1 | NP_001240660.1 | 13 | 1668 | ||
NM_153414.4 | NP_700463.2 | 13 | 1636 | |||
GenBank mRNA | AK038979.1 | - | 13 | 1369 | ||
AK077634.1 | - | 13 | 1637 | |||
AK136992.1 | - | 13 | 1363 | |||
AK150195.1 | - | 13 | 1363 | |||
BC028953.1 | - | 12 | 1326 | 5362016 | ||
BC055700.1 | - | 13 | 1335 | 6808816 |