Search
Search

Arl14ep| Gene Symbol: Arl14ep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025102.1 | NP_001020273.1 | 3 | 666 | ||
| NM_173750.2 | NP_776111.1 | 3 | 620 | |||
| GenBank mRNA | AK076781.1 | - | 3 | 621 | ||
| AK134965.1 | - | 3 | 657 | |||
| BC054106.1 | - | 3 | 652 | 30021622 | ||