Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145743.2 | NP_665686.2 | 6 | 836 | ||
GenBank mRNA | AF397909.1 | - | 5 | 539 | ||
AF520417.1 | - | 6 | 685 | |||
AK044595.1 | - | 6 | 712 | |||
AK170996.1 | - | 6 | 687 | |||
BC023308.1 | - | 5 | 513 | |||
BC089595.1 | - | 6 | 686 | 30118411 |