Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Plk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183152.3 | NP_898975.2 | 8, 9 | 1081 | ||
GenBank mRNA | AK031968.1 | - | 8, 9 | 1082 | ||
BC086684.1 | - | 3, 4 | 378 | 30679265 | ||
BC099679.1 | - | 8, 9 | 1050 | 6489647 |