Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pacs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081170.1 | NP_001074639.1 | 10 | 1060 | ||
NM_001291444.1 | NP_001278373.1 | 10 | 1060 | |||
NM_001291445.1 | NP_001278374.1 | 9 | 970 | |||
GenBank mRNA | AK030003.1 | - | 5 | 428 | ||
AK036581.1 | - | 9 | 946 | |||
AK054000.1 | - | 10 | 1035 | |||
AK122326.1 | - | 8 | 749 |