Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfb1i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289550.1 | NP_001276479.1 | 5 | 527 | ||
NM_001289551.1 | NP_001276480.1 | 5 | 527 | |||
NM_001289552.1 | NP_001276481.1 | 4 | 384 | |||
NM_001289553.1 | NP_001276482.1 | 4 | 411 | |||
GenBank mRNA | AK003170.1 | - | 5 | 362 | ||
AK041233.1 | - | 5 | 355 | |||
AK085042.1 | - | 4 | 253 | |||
AK087114.1 | - | 5 | 362 | |||
AK156423.1 | - | 5 | 362 | |||
AK158409.1 | - | 4 | 236 | |||
BC002049.1 | - | 5 | 361 | 3489459 | ||
BC056362.1 | - | 4 | 1636 | 6816930 | ||
DQ143891.1 | - | 4 | 412 | |||
DQ143892.1 | - | 3 | 227 | |||
DQ143893.1 | - | 3 | 205 | |||
DQ143894.1 | - | 3 | 200 | |||
DQ143895.1 | - | 3 | 616 | |||
DQ143897.1 | - | 3 | 655 | |||
DQ143898.1 | - | 4 | 489 | |||
DQ143899.1 | - | 3 | 616 | |||
DQ143900.1 | - | 4 | 424 | |||
L22482.1 | - | 4 | 488 |