Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TH SIRNA Assays ›
Gene Symbol:
Th
Gene Name:
tyrosine hydroxylase
Gene Aliases:
-
Chromosome Location:
Chr.7: 142892776 - 142899966 on Build GRCm38
UniGene ID:
Mm.1292
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS278329
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Contact Us
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Th
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_009377.1
NP_033403.1
11
1182
GenBank mRNA
AK139568.1
-
1399
M69200.1
-
11
1182
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
synaptic transmission, dopaminergic
dopamine biosynthetic process from tyrosine
heart development
visual perception
learning
memory
mating behavior
locomotory behavior
regulation of heart contraction
aromatic amino acid family metabolic process
organ morphogenesis
aminergic neurotransmitter loading into synaptic vesicle
neurotransmitter biosynthetic process
dopamine biosynthetic process
epinephrine biosynthetic process
norepinephrine biosynthetic process
catecholamine biosynthetic process
eye photoreceptor cell development
eating behavior
response to ethanol
embryonic camera-type eye morphogenesis
oxidation-reduction process
monooxygenase activity
tyrosine 3-monooxygenase activity
iron ion binding
ferrous iron binding
ferric iron binding
oxidoreductase activity
amino acid binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced pteridine as one donor, and incorporation of one atom of oxygen
oxygen binding
enzyme binding
protein domain specific binding
tetrahydrobiopterin binding
dopamine binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0