Search
Search

4930474N05Rik| Gene Symbol: 4930474N05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175008.3 | NP_778173.1 | 3 | 394 | ||
| GenBank mRNA | AK076810.1 | - | 3 | 395 | ||
| AK132960.1 | - | 403 | ||||
| BC096580.1 | - | 3 | 451 | 3470874 | ||
| BC132442.1 | - | 3 | 390 | 40130719 | ||
| BC132446.1 | - | 3 | 390 | 40130723 | ||
| BC145191.1 | - | 3 | 361 | |||