Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nr1h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285517.1 | NP_001272446.1 | 4 | 380 | ||
NM_001285518.1 | NP_001272447.1 | 4 | 383 | |||
NM_001285519.1 | NP_001272448.1 | 4 | 380 | |||
NM_009473.3 | NP_033499.1 | 4 | 387 | |||
GenBank mRNA | AK054508.1 | - | 4 | 367 | ||
AK077620.1 | - | 4 | 362 | |||
AK134489.1 | - | 4 | 342 | |||
AK156777.1 | - | 4 | 364 | |||
BC066025.1 | - | 4 | 338 | 6829527 | ||
BC106148.1 | - | 3 | 116 | |||
BQ573162.1 | - | 4 | 357 | |||
EU869275.2 | - | 2 | 105 | |||
U09419.1 | - | 3 | 205 |