Search
Search

Morn4| Gene Symbol: Morn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198108.2 | NP_932776.1 | 5 | 511 | ||
| GenBank mRNA | AK154147.1 | - | 5 | 499 | ||
| AK159230.1 | - | 5 | 498 | |||
| BC023055.1 | - | 5 | 477 | |||
| BC057060.1 | - | 4 | 887 | |||
| CV563048.1 | - | 5 | 437 | |||