Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110791.1 | NP_001104261.1 | 7, 8 | 1171 | ||
NM_011750.2 | NP_035880.2 | 7, 8 | 1171 | |||
GenBank mRNA | AK133895.1 | - | 7, 8 | 960 | ||
AK146151.1 | - | 7, 8 | 932 | |||
AK147081.1 | - | 7, 8 | 940 | |||
AK152914.1 | - | 6, 7 | 873 | |||
AK154705.1 | - | 7, 8 | 939 | |||
AK157845.1 | - | 7, 8 | 940 | |||
AK195027.1 | - | 1, 2 | 69 | |||
BC009091.1 | - | 7, 8 | 916 | 3157495 | ||
BC055370.1 | - | 7, 8 | 930 | 30055109 | ||
CD563870.1 | - | 1, 2 | 32 | |||
X85802.1 | - | 7, 8 | 928 |