Search
Search

Siglech| Gene Symbol: Siglech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178706.4 | NP_848821.2 | 2 | 668 | ||
| GenBank mRNA | AK032377.1 | - | 2 | 621 | ||
| AK045719.1 | - | 3 | 298 | |||
| AK047813.1 | - | 2 | 585 | |||
| AK139362.1 | - | 3 | 569 | |||
| BC098229.1 | - | 2 | 607 | 1430130 | ||
| BC120501.1 | - | 2 | 610 | 40129424 | ||
| BC144742.1 | - | 2 | 610 | 9053290 | ||
| BC144743.1 | - | 2 | 610 | 9053291 | ||
| DQ157764.1 | - | 2 | 587 | |||
| DQ208935.1 | - | 2 | 613 | |||